missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98690-50ul
This item is not returnable.
View return policy
Description
ARID3B Polyclonal antibody specifically detects ARID3B in Human samples. It is validated for Western Blot
Specifications
| ARID3B | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| ARID domain-containing protein 3B, AT rich interactive domain 3B (BRIGHT- like), AT rich interactive domain 3B (BRIGHT-like), AT-rich interactive domain-containing protein 3B, BDP, bright and dead ringer protein, DRIL2 | |
| Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human ARID3B. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10620 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction