missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | ARID2 |
|---|---|
| Concentration | 233 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236039
|
Proteintech
23406-1-AP-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234455
|
Proteintech
23406-1-AP-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ARID2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARID2. This antibody recognizes human, mouse antigen. The ARID2 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| ARID2 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| BAF200; DKFZp686G052; DKFZp779P0222; FLJ30619; KIAA1557; p200 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 233 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 196528 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title