missing translation for 'onlineSavingsMsg'
Learn More

ARID1A Antibody (CL3595) [DyLight 350], Novus Biologicals™

Product Code. 30300780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30300780 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30300780 Supplier Novus Biologicals Supplier No. NBP261623UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARID1A Monoclonal antibody specifically detects ARID1A in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID1A
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL3595
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ARID domain-containing protein 1A, AT rich interactive domain 1A (SWI- like), AT rich interactive domain 1A (SWI-like), AT-rich interactive domain-containing protein 1A, B120SWI-like protein, BAF250a, BAF250SWI/SNF complex protein p270, BM029, brain protein 120, BRG1-associated factor 250, BRG1-associated factor 250a, C10rf4, C1orf4SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatinsubfamily F member 1, chromatin remodeling factor p250, hELD, hOSA1, matrix associated, actin dependent regulator of chromatin, Osa homolog 1, OSA1, OSA1 nuclear protein, P270, SMARCF1, subfamily f, member 1
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from O14497, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8289
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.