missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ARHGAP44 Polyclonal antibody specifically detects ARHGAP44 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ARHGAP44 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:250-1:500, ELISA 1:10000, Immunohistochemistry 1:50-1:150, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:50-1:150 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | Rho GTPase activating protein 44, rho GTPase-activating protein RICH2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to N terminal amino acids 1-60 of Mouse RICH2. Sequence: MKKQFNRMRQ LANQTVGRA E KTEVLSED LL QVEKRLE LVK QVSHST HKKL TACLQ GQQGA |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?