missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92692-0.02ml
This item is not returnable.
View return policy
Description
ARHGAP1 Polyclonal antibody specifically detects ARHGAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ARHGAP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:100 | |
| CDC42 GTPase-activating protein, CDC42GAPp50-RhoGAP, GTPase-activating protein rhoOGAP, p50rhoGAP, Rho GTPase activating protein 1, rho GTPase-activating protein 1, RHOGAP, RHOGAP1Cdc42GAP, Rho-related small GTPase protein activator, Rho-type GTPase-activating protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 210-439 of human ARHGAP1 (NP_004299.1). VLKYDDFLKSTQKSPATAPKPMPPRPPLPNQQFGVSLQHLQEKNPEQEPIPIVLRETVAYLQAHALTTEGIFRRSANTQVVREVQQKYNMGLPVDFDQYNELHLPAVILKTFLRELPEPLLTFDLYPHVVGFLNIDESQRVPATLQVLQTLPEENYQVLRFLTAFLVQISAHSDQNKMTNTNLAVVFGPNLLWAKDAAITLKAINPINTFTKFLLDHQGELFPSPDPSGL | |
| 0.02 mL | |
| GPCR, Signal Transduction | |
| 392 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction