missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ARF7 Recombinant Protein

Brand:  Abnova™ H00080117-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104773

  • 528.00€ / 25µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH34354
Solution
80117
FLJ22595 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
ARF7/FLJ22595
ARL14
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
46.86
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARL14
Human
Yes
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers