missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Mouse anti-Human, Mouse, Rat, Clone: 1A9/5, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | ARF1 |
|---|---|
| Clone | 1A9/5 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
ARF1 Monoclonal specifically detects ARF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| ARF1 | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| ADP-ribosylation factor 1 | |
| C-terminal human ARF1 peptide SNQLRNQ | |
| Primary | |
| Store at 4°C. |
| 1A9/5 | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 375 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title