missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Antibody (1B3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97935FR
This item is not returnable.
View return policy
Description
ARF1 Monoclonal specifically detects ARF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ARF1 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ARF1 | |
| Recombinant human ARF1 (1-181aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Anti-human ARF1 mAb, clone 1B3, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ARF1 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1B3 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| ADP-ribosylation factor 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 375 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction