Learn More
Aquaporin-4 Antibody (C9401) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43559
This item is not returnable.
View return policy
Description
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Neutralization
Specifications
| Aquaporin-4 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunoprecipitation, Neutralization | |
| aquaporin 4, MGC22454 | |
| The original antibody was generated by immunizing an aquaporin 4 knock out mice with human AQP4 as an immunogen expressed on the surface of baculovirus. (Uniprot# P55087) | |
| 0.2 mg | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 361 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA, Immunoprecipitation, Neutralization | |
| C9401 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |