Learn More
AQP8 Monoclonal Antibody (OTI6E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810725S
This item is not returnable.
View return policy
Description
AQP8 Monoclonal antibody specifically detects AQP8 in Human samples. It is validated for Western BlotSpecifications
| AQP8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P60568 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3558 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6E6 | |
| Unconjugated | |
| IL2 | |
| AI255744; AQP8; AQP-8; aquaporin 8; Aquaporin8; aquaporin-8; membrane water channel; MGC93492; water channel | |
| Human recombinant protein fragment corresponding to amino acids 21-153 of human IL2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.