missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AQP8 Monoclonal Antibody (OTI6E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810725
This item is not returnable.
View return policy
Description
Aquaporin-8 is a protein that in humans is encoded by the AQP8 gene. Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. Forms a water-specific channel; mercury-sensitive. Not permeable to glycerol or urea. Expressed only in pancreas and colon.Specifications
| AQP8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O94778 | |
| AQP8 | |
| Full length human recombit protein of human AQP8 produced in HEK293T. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6E6 | |
| Unconjugated | |
| AQP8 | |
| AQP-8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 343 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction