missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L1 Antibody (CL0173) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52871AF594
This item is not returnable.
View return policy
Description
Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human samples. It is validated for Western Blot
Specifications
| Apolipoprotein L1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4 | |
| This antibody was developed using a recombinant protein derived from O14791, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology | |
| 8542 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL0173 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction