missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein B/ApoB Antibody (6G6), Novus Biologicals™

Mouse Monoclonal Antibody

196.00€ - 590.00€

Specifications

Antigen Apolipoprotein B/ApoB
Clone 6G6
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18624036
View Documents
Novus Biologicals
NBP2-37598-0.025ml
0.025 mL
196.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18165458
View Documents
Novus Biologicals
NBP2-37598
0.1 mL
590.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Apolipoprotein B/ApoB Monoclonal specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Apolipoprotein B/ApoB
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000
Monoclonal
Ascites
RUO
Human
Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100
APOB
IgG1
Unpurified
6G6
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Ascites with 0.03% Sodium Azide
338
Purified recombinant fragment of human ApoB expressed in E. Coli.
Primary
516 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.