missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A-II/ApoA2 Rabbit anti-Human, Mouse, Rat, Clone: 5F7U1, Novus Biologicals™
Rabbit Monoclonal Antibody
201.00€ - 497.00€
Specifications
| Antigen | Apolipoprotein A-II/ApoA2 |
|---|---|
| Clone | 5F7U1 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18384942
|
Novus Biologicals
NBP3-16168-20UL |
20 μg |
201.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18373202
|
Novus Biologicals
NBP3-16168-100UL |
100 μg |
497.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein A-II/ApoA2 Monoclonal antibody specifically detects Apolipoprotein A-II/ApoA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| Apolipoprotein A-II/ApoA2 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APOA2, apoAII, Apo-AII, ApoA-II, Apolipoprotein A2, apolipoprotein A-II | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human Apolipoprotein A-II/ApoA2 (P02652). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 5F7U1 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Lipid and Metabolism | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 336 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title