missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APJ/Apelin receptor Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS63
This item is not returnable.
View return policy
Description
APJ/Apelin receptor Polyclonal antibody specifically detects APJ/Apelin receptor in Human, Equine, Hamster samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot (Negative)
Specifications
| APJ/Apelin receptor | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Equine, Hamster | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 4-7 μg/mL, Western Blot (Negative) | |
| AGTRL1HG11, angiotensin II receptor-like 1, Angiotensin receptor-like 1, apelin receptor, APJ (apelin) receptor, APJ receptor, APJFLJ96609, APJR, FLJ90771, G protein-coupled receptor APJ, G-protein coupled receptor APJ, G-protein coupled receptor HG11, HG11 orphan receptor, MGC45246 | |
| Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human APJ/Apelin receptor. | |
| 0.05 mL | |
| GPCR | |
| 187 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction