missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apelin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12282
This item is not returnable.
View return policy
Description
Apelin Polyclonal antibody specifically detects Apelin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Apelin | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:50000, Immunohistochemistry 1:50-1:150, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:50-1:150 | |
| AGTRL1 ligand, APEL, apelin, apelin, AGTRL1 ligand, APJ endogenous ligand, XNPEP2 | |
| Synthetic peptide taken within amino acid region 25-77 on human Apelin protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| 8862 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction