missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
302.00€ - 443.00€
Specifications
| Antigen | APE |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Research Discipline | Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656561
|
Novus Biologicals
NBP2-76495-25ul |
25 μL |
302.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18390611
|
Novus Biologicals
NBP2-76495 |
100 μL |
443.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APE Polyclonal specifically detects APE in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| APE | |
| Unconjugated | |
| Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease | |
| APEX1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Polyclonal | |
| Rabbit | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 328.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: DKEGYSGVGLLSRQCPLKVSYGIGEEEHDQEGRVIVAEFDSFVLVTAYVPNAGRGLVRLEYRQRWDEAFRKFLKGLASRKPLVLCGDLNVAHEEIDLRNPKGNKKNAGFTPQEGAPHR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title