missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€
Specifications
| Antigen | Apc7 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
Apc7 Polyclonal specifically detects Apc7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Apc7 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| NP_057322 | |
| 51434 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human APC7. The immunogen is located within amino acids 520 - 570 of APC7. Amino Acid Squence: LDPNDQKSLEGMQKMEME | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| anaphase promoting complex subunit 7, anaphase-promoting complex subunit 7, APC7Cyclosome subunit 7 | |
| ANAPC7 | |
| IgG | |
| Affinity Purified | |
| 68 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title