missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ AP2S1 (Human) Recombinant Protein

Human AP2S1 full-length ORF ( AAH06337, 1 a.a. - 142 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00001175-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16103271

  • 349.00€ / 10µg

Estimated Shipment: 12-07-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MIRFILIQNRAGKTRLAKWYMQFDDDEKQKLIEEVHAVVTVRDAKHTNFVEFRNFKIIYRRYAGLYFCICVDVNDNNLAYLEAIHNFVEVLNEYFHNVCELDLVFNFYKVYTVVDEMFLAGEIRETSQTKVLKQLLMLQSLE
Specifications

Specifications

AAH06337
adaptor-related protein complex 2, sigma 1 subunit
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AP2S1
GST
1175
Wheat germ expression system
10 μg
AP17, AP17-DELTA, CLAPS2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers