Learn More
Fisherbrand™ AP Bio 400 Air Purification Unit
Product Code. 17408982 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Airflow Rate:
100 to 400 m3/h
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
17408982 100 to 400 m3/h Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17408982
  • ONLINE EXCLUSIVE PRICE
    2105.25€
    1570.03€ / Pack of 1
    Save 535.22€
    25% Off
 Restricted Item
Estimated Shipment: 30-11-2026
The unique AP BIO 400 Air Purifier is the best and proven way to combat airborne transmission of viruses and bacteria in almost any closed environment.
  • Unique Technology - AP BIO is the only bio decontaminating solution equipped with ePM1 50% (F7) and HEPA H14 filters with AirFend™ technology which retains and deactivates bacteria and viruses (including Sars-CoV-2 responsible for Covid-19).
  • Scientifically Tested - The AirFend™ solution applied to the filters of the AP BIO Air Purifier has a virucidal action, including on the H1N1 influenza virus and the coronavirus (SARS-CoV-2) according to the test protocol standard EN 14476.
  • With the unique AP BIO 400 Air Purifiers from FläktGroup with the revolutionary AirFend™ filtration technology, viruses and bacteria are nearly 100% deactivated – including Sars-CoV-2 – when air passes through the filters in the unit.
  • AirFend™ is a 100% natural solution.
  • The AP Bio is the perfect choice for healthy and safe indoor air in any area such as homes, hospitals, schools, offices etc.
  • The air purifiers known on the market are generally effective in filtering and improving the overall air quality and can be installed in rooms where filtration is usually not possible, but they do not protect 100% against viruses. Indeed, thanks to a high air renewal rate and high filtration, viruses are captured and filtered but they remain active.
  • The unit also purifies the air of dust and other particles with its 50% ePM1 (F7) and HEPA H14 filters.
  • AP BIO air purifier uses an active air treatment process, unlike other passive air treatment solutions.
  • AP BIO is a mobile unit which can easily be moved to different rooms and used in a very flexible way.
  • The unit is easy and quick to install and gives easy access for maintenance of filters without tools.
  • Epoxy painted and galvanized steel casing
  • 2 AirFend™ mini-fold filters ePM1 50% (F7), 2 AirFend™ HEPA H14 absolute filters (model AP BIO 2400)
  • Flow rate adjustment control panel
  • Indicator light for filter wear rate  
  • EC-motor fan with low energy consumption
  • Silencer (model AP BIO 2400)
  • Perforated air intake grille with low pressure drop
  • Easy push-pull grid opening and closing
  • Tool-less clamping system
  • Waterproof panel wall
  • Mobile unit on casters
  • Duration of filter effectiveness: 3 months in a highly contaminated environment (hospital), and up to 6 months in a normal environment (hotels, offices, schools).
Specifications
Airflow Rate 100 to 400 m3/h
Weight (English) 81.6 lb.
Weight (Metric) 37 Kg
Depth (English) 1 ft. 5 1/8 in.
Width (English) 1 ft. 5.125 in.
Height (Metric) 840 mm
Depth (Metric) 435 mm
Width (Metric) 435 mm
Product Type Air Purifier
Voltage 230 V