missing translation for 'onlineSavingsMsg'
Learn More

zinc finger protein 397, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16144729
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16144729 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16144729 Supplier Abnova Supplier No. H00084307D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against a full-length human ZNF397 protein.

Sequence: MAVESGVISTLIPQDPPEQELILVKVEDNFSWDEKFKQNGSTQSCQELFRQQFRKFCYQETPGPREALSRLQELCYQWLMPELHTKEQILELLVLEQFLSILPEELQIWVQQHNPESGEEAVTLLEDLEREFDDPGQQVPASPQGPAVPWKDLTCLRASQESTDIHLQPLKTQLKSWKPCLSPKSDCENSETATKEGISEEKSQGLPQEPSFRGIKLSRPPKASSAIRWECVSPGSFPGDIIAAEATHSTISCFAINTLPATILPSKNVNRKYFS

Specifications

Antigen zinc finger protein 397
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ZNF397 protein.
Formulation PBS with no preservative; pH 7.4
Gene ZNF397
Gene Accession No. NM_032347.1
Gene Alias MGC13250/ZNF47/ZSCAN15
Gene Symbols ZNF397
Host Species Rabbit
Immunogen ZNF397 (NP_115723.1, 1 a.a. ∼ 275 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 84307
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.