Learn More
Zap70 (pY292) Mouse anti-Human, PE, Clone: J34-602, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558510
Description
The 70kDa ζ chain-associated protein (ZAP70) is a Syk-family protein tyrosine kinase (PTK) that associates with the ζ subunit of the T cell antigen receptor (TCR) and undergoes tyrosine phosphorylation following TCR stimulation. ZAP70 contains two SH2-like domains with the PTK domain located at the C-terminus. TCR-mediated Lck activity leads to phosphorylation of ZAP70 on Tyrosine 493 in the regulatory loop of the PTK domain leading to upregulation of ZAP70 kinase activity. Tyrosine 292 (Y292), in the linker region between the SH2 and PTK domains, is autophosphorylated by the activated PTK domain. By binding with c-Cbl, the phosphorylated Y292 can down-regulate TCR signaling. The J34-602 antibody recognizes ZAP70 phosphorylated at Y292.
Intracellular Staining
Specifications
| Zap70 (pY292) | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Phosphorylated Human ZAP70 Peptide | |
| 50 Tests | |
| Cell Biology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| J34-602 | |
| ZAP-70 (pY292) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.