missing translation for 'onlineSavingsMsg'
Learn More

anti-WARS Matched Antibody Pair, Abnova™

Product Code. 16182784
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16182784 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16182784 Supplier Abnova Supplier No. H00007453AP51.1Set
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Capture antibody: mouse monoclonal anti-WARS (100μg); Detection antibody: rabbit purified polyclonal anti-WARS (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human WARS

Host Species: Mouse, Rabbit
Species Reactivity: Hu

ELISA Pair

Spécification

Antigen WARS
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human WARS.
Gene WARS
Gene Alias GAMMA-2/IFI53/IFP53
Gene Symbols WARS
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7453
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-WARS (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-WARS (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Purified
Afficher plus Afficher moins
Nom du produit
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.