missing translation for 'onlineSavingsMsg'
Learn More

Villin Mouse anti-Human, Unlabeled, Clone: 12, BD

Product Code. 15825419
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15825419 50 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15825419 Supplier BD Biosciences Supplier No. 610358
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The isolated intestinal microvillus cytoskeleton (core) consists of four major proteins: actin, villin, fimbrin, and brush border myosin-I. These proteins can assemble in vitro into structures resembling native microvillus cores. Of these components, villin, and brush border myosin-I show tissue-specific expression, so they may be involved in the morphogenesis of intestinal microvilli. Found in association with the microvillar actin bundles of the intestinal brush border, villin is a 95kDa protein composed of two very similar domains of approximately 44kDa each, the core, and a C-terminal domain of 8kDa, the headpiece. The core has been shown to contain villin's Ca2+ regulated capping, nucleating, and severing activities, but it cannot induce the formation of microfilament bundles without the headpiece. Villin is a useful differentiation marker of early embryogenesis and may be useful in diagnosis and follow-up of colorectal cancers. It has been demonstrated that villin is necessary for both the cytoskeletal and membrane protein organization of a functional brush border.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Villin
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 12
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.