missing translation for 'onlineSavingsMsg'
Learn More

Anti-Total OXPHOS Antibody Cocktail antibody [RM1134], Abcam

Product Code. 32095749 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
Unit Size:
100µL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32095749 1 mL 1mL
32094988 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32095749 Supplier Abcam Supplier No. ab3172701mL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Polyclonal Antibody

Rabbit Recombinant Multiclonal NDUFB8 antibody. Suitable for ICC/IF, Flow Cyt (Intra), WB, IHC-P and reacts with Human samples. Cited in 1 publication.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen NDUFB8
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Western Blot
Cellular Localization Mitochondrion inner membrane; Single-pass membrane protein; Matrix side
Classification Recombinant Polyclonal
Clone RM1134
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene NDUFB8
Gene Accession No. O95169
Host Species Rabbit
Immunogen NDUFB8
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cardiovascular
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene NDUFB8
Target Molecule NDUFB8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.