missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFSF18, Human, Matched Antibody Pair (AP51), Abnova™
Brand: Abnova H00008995-AP51.1Set
Includes: Capture antibody: mouse monoclonal anti-TNFSF18 (100μg); Detection antibody: rabbit purified polyclonal anti-TNFSF18 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFSF18
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair
Specifications
| TNFSF18 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFSF18. | |
| AITRL/GITRL/MGC138237/TL6/hGITRL | |
| Mouse, Rabbit | |
| RUO | |
| 8995 | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| TNFSF18 | |
| TNFSF18 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TNFSF18 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-TNFSF18 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto