Learn More
anti-TNFSF15, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand: Abnova H00009966-AP22.1Set
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-TNFSF15 (100μg) , Detection antibody: mouse purified polyclonal anti-TNFSF15 (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
| TNFSF15 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFSF15. | |
| MGC129934/MGC129935/TL1/TL1A/VEGI/VEGI192A | |
| Mouse, Rabbit | |
| RUO | |
| 9966 | |
| Human | |
| Liquid |
| ELISA | |
| Unconjugated | |
| TNFSF15 | |
| TNFSF15 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TNFSF15 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-TNFSF15 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |