Learn More
TNFRSF6B, Human, Matched Antibody Pair (AP41), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00008771-AP41.1Set
Product Code. 16102884
  • 843.00€ / 1 set
Includes: Capture antibody: mouse monoclonal anti-TNFRSF6B (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TNFRSF6B, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
TNFRSF6B
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFRSF6B.
DCR3/DJ583P15.1.1/M68/TR6
Mouse
RUO
8771
Human
Liquid
ELISA
Biotin
TNFRSF6B
TNFRSF6B
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TNFRSF6B (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TNFRSF6B, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.