Learn More
anti-TNFRSF1B Matched Antibody Pair, Abnova™
Description
Includes
Capture antibody: rabbit MaxPab affinity purified polyclonal anti-TNFRSF1B (100μg); Detection antibody: mouse monoclonal anti-TNFRSF1B, IgG2a κ (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFRSF1B
Host Species: Rabbit, Mouse
Isotype: IgG, IgG2a κ
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
Specifications
| Antigen | TNFRSF1B |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG, IgG2a κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.