Learn More
TNFRSF10B, Human, IP-WB Antibody Pair (PW2), Abnova™
Product Code. 16142894
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16142894 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16142894 Supplier Abnova Supplier No. H00008795PW2.1Set
Includes
Antibody pair for IP: mouse monoclonal anti-TNFRSF10B (300μg), Antibody pair for WB: rabbit polyclonal anti-TNFRSF10B (50μL)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen TNFRSF10B
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene TNFRSF10B
Gene Alias CD262/DR5/KILLER/KILLER/DR5/TRAIL-R2/TRAILR2/TRICK2/TRICK2A/TRICK2B/TRICKB/ZTNFR9
Gene Symbols TNFRSF10B
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8795
Includes Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-TNFRSF10B (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-TNFRSF10B (50 ul)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid