Learn More
TNF Rat anti-Mouse, FITC, Clone: MP6-XT22, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554418
Additional Details : Weight : 0.01000kg
Description
The MP6-XT22 antibody reacts with mouse tumor necrosis factor (TNF, also known as TNF-α). The immunogen used to generate this hybridoma was recombinant mouse TNF.
Host Species: Rat
Clone: MP6-XT22
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Recombinant mouse TNF
Intracellular Staining
Specifications
TNF | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant mouse TNF | |
0.1 mg | |
Immunology | |
Mouse | |
IgG1 |
Flow Cytometry | |
MP6-XT22 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.