Learn More
TNF Rat anti-Mouse, FITC, Clone: MP6-XT22, BD
Rat Monoclonal Antibody
126.00€ - 313.00€
Specifications
Antigen | TNF |
---|---|
Clone | MP6-XT22 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15820739
|
BD Biosciences
561064 |
25 μg |
126.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
15807688
|
BD Biosciences
554418 |
0.1 mg |
313.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
The MP6-XT22 antibody specifically binds to mouse Tumor Necrosis Factor (TNF, also known as TNF-α). TNF is produced by many activated cell types including monocytes, macrophages, astrocytes, granulocytes, mast cells, T and B lymphocytes, NK cells, keratinocytes, fibroblasts, adipocytes, and certain tumor cells. Activated cells express type II transmembrane TNF glycoproteins that associate as homotrimeric complexes. After enzymatic cleavage, the extracellular regions of membrane TNF are shed as soluble homotrimers. TNF is a potent multifunctional cytokine that can exert regulatory and cytotoxic effects on a wide range of normal lymphoid and non-lymphoid cells and tumor cells. Although TNF serves as a primary mediator in protective immune responses against microbial and viral pathogens, it can also drive systemic pathophysiologic responses including septic shock, cachexia and autoimmune diseases. Mouse TNF exerts its biological activities by binding and signaling through cell surface membrane Type I and Type II TNF Receptors (aka, TNFRI/CD120a and TNFRII/CD120b, respectively).
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
TNF | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
IgG1 | |
Affinity Purified |
MP6-XT22 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.