missing translation for 'onlineSavingsMsg'
Learn More

Anti-TGIF antibody [EP637Y], Abcam

Product Code. 32163327 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
Unit Size:
100µL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32163327 1 mL 1mL
32159392 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32163327 Supplier Abcam Supplier No. ab529551mL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Rabbit Recombinant Monoclonal TGIF antibody. Suitable for IP, WB and reacts with Human samples. Cited in 10 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen TGIF1 (TGIF)
Applications Immunoprecipitation, Western Blot
Cellular Localization Nucleus
Classification Recombinant Monoclonal
Clone EP637Y
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene TGIF1
Gene Accession No. Q15583
Host Species Rabbit
Immunogen TGIF1 (TGIF)
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Oncology
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Conditional Ambient
Target Gene TGIF1
Target Molecule TGIF1 (TGIF)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.