Learn More
anti-STAM, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00008027-PW1.1Set
Product Code. 16172814
  • 843.00€ / 1 set
Includes: Antibody pair for IP: mouse monoclonal anti-STAM (300μg), Antibody pair for WB: mouse monoclonal anti-STAM (50μg)
Specifications
STAM
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
DKFZp686J2352/STAM1
Mouse
RUO
8027
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
STAM
STAM
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-STAM (300 ug) 2. Antibody pair for WB: mouse monoclonal anti-STAM (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.