missing translation for 'onlineSavingsMsg'
Learn More

Anti-SPOCK1 antibody, Abcam

Product Code. 32100227 Shop All Abcam Products
Change view
Click to view available options
Quantity:
50 μL
50 μg
Unit Size:
50µL
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32100227 50 μg 50µg
32097543 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32100227 Supplier Abcam Supplier No. ab22993550ug
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rabbit Polyclonal SPOCK1 antibody. Suitable for WB, IHC-P and reacts with Mouse, Human samples. Cited in 15 publications. Immunogen corresponding to Recombinant Fragment Protein within Human SPOCK1 aa 250 to C-terminus.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen SPOCK1
Applications Immunohistochemistry (FFPE), Western Blot
Cellular Localization Secreted; Extracellular space; Extracellular matrix
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.03% Proclin 300, Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene SPOCK1
Gene Accession No. Q08629
Host Species Rabbit
Immunogen SPOCK1
Purification Method Affinity purification
Quantity 50 μg
Regulatory Status RUO
Research Discipline Oncology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene SPOCK1
Target Molecule SPOCK1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.