Learn More
anti-SPEG Matched Antibody Pair, Abnova
Brand: Abnova H00010290-AP11.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-SPEG (100μg); Mouse monoclonal anti-SPEG, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Expression of this gene is thought to serve as a marker for differentiated vascular smooth muscle cells which may have a role in regulating growth and differentiation of this cell type. The encoded protein is highly similar to the corresponding rat and mouse proteins. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of only one variant has been defined.
ELISA
Specifications
| SPEG | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SPEG. | |
| APEG1/BPEG/KIAA1297/MGC12676/SPEGalpha/SPEGbeta | |
| Mouse, Rabbit | |
| 1 Set | |
| Cell Cycle | |
| 10290 | |
| Human | |
| IgG1 |
| ELISA | |
| Unconjugated | |
| SPEG | |
| SPEG | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit affinity purified polyclonal anti-SPEG (100 ug) 2. Detection antibody: mouse monoclonal anti-SPEG, IgG1 Kappa (20 ug) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.