Learn More
anti-SORD, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00006652-PW1.1Set
Product Code. 16089398
  • 843.00€ / 1 set
Includes: Antibody pair for IP: mouse monoclonal anti-SORD (300μg), Antibody pair for WB: rabbit polyclonal anti-SORD (50μL)
Specifications
SORD
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
SORD1
Mouse, Rabbit
RUO
6652
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
SORD
SORD
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-SORD (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-SORD (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.