missing translation for 'onlineSavingsMsg'
Learn More

SLA-DR Mouse anti-Pig, Unlabeled, Clone: 1053H2-18, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 553642

Product Code. 15807318

  • ONLINE EXCLUSIVE PRICE
    237.00€
    236.48€ / Each
    Save 0.52€
    0.22% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 1053h2-18-1 antibody reacts with swine leukocyte antigen-DR (SLA-DR) expressed on subsets of pig B and T lymphocytes and monocytes, but not on granulocytes. It also reacts with endothelial cells, Langerhans cells, thymocytes, and macrophages. An interesting observation is that SLA-DR is preferentially expressed on CD8+ T cells.

Host Species: Mouse
Clone: 1053H2-18
Isotype: IgG2a κ
Species Reactivity: Pig
Immunogen: Mouse L cells transfected with pig SLA-DR gene

Flow Cytometry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLA-DR
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse L cells transfected with pig SLA-DR gene
0.1 mg
Immunology
Pig
IgG2a κ
Flow Cytometry
1053H2-18
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.