missing translation for 'onlineSavingsMsg'
Learn More

anti-SERPINF1 Matched Antibody Pair, Abnova

Product Code. 16102424
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16102424 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16102424 Supplier Abnova Supplier No. H00005176AP31.1Set
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Rabbit MaxPab™ affinity purified polyclonal anti-SERPINF1 (100μg); Biotinylated rabbit MaxPab™ affinity purified polyclonal anti-SERPINF1 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.

The PEDF gene is a member of the serpin gene family. Serpins are a group of serine protease inhibitors, some of which have also been reported to exhibit neurotrophic activity.

ELISA

Specifications

Antigen SERPINF1
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Biotin
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SERPINF1.
Gene SERPINF1
Gene Alias EPC-1/PEDF/PIG35
Gene Symbols SERPINF1
Host Species Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5176
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-SERPINF1 (100 μg) 2. Detection antibody: biotinylated rabbit MaxPab™ affinity purified polyclonal anti-SERPINF1 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.