Learn More
SENP8, Human, IP-WB Antibody Pair (PW3), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00123228-PW3.1Set
Includes: Antibody pair for IP: rabbit polyclonal anti-SENP8 (300μL), Antibody pair for WB: mouse monoclonal anti-SENP8 (50μg)
Specifications
| SENP8 | |
| Monoclonal/Polyclonal | |
| This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
| DEN1/HsT17512/NEDP1/PRSC2 | |
| Mouse, Rabbit | |
| RUO | |
| 123228 | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| SENP8 | |
| SENP8 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-SENP8 (300 ul) 2. Antibody pair for WB: mouse monoclonal anti-SENP8 (50 ug) | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |