missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-SDSL Ab-1 Matched Antibody Pair, Abnova™
Description
Includes
Capture antibody: mouse monoclonal anti-SDSL (100μg); Detection antibody: rabbit MaxPab™ affinity purified polyclonal anti-SDSL (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Host Species: Mouse, Rabbit
Cross Reactivity: Human
ELISA Pair
Specifications
Specifications
| Antigen | SDSL |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SDSL. |
| Gene | SDSL |
| Gene Alias | SDS-RS1 |
| Gene Symbols | SDSL |
| Host Species | Mouse, Rabbit |
| Quantity | 1 Set |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?