missing translation for 'onlineSavingsMsg'
Learn More

anti-RICTOR, Human, IP-WB Antibody Pair, Abnova™

Product Code. 16113914
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16113914 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16113914 Supplier Abnova Supplier No. H00253260PW1.1Set
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Antibody pair for IP: mouse monoclonal anti-RICTOR (300μg), Antibody pair for WB: rabbit polyclonal anti-RICTOR (50μL)

For immunoprecipitation and to detect the precipitated protein in western blot.

Specifications

Antigen RICTOR
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene RICTOR
Gene Alias DKFZp686B11164/KIAA1999/MGC39830/mAVO3
Gene Symbols RICTOR
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253260
Includes Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-RICTOR (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-RICTOR (50 ul)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.