Learn More
anti-RFXANK, Human, Matched Antibody Pair, Abnova™
Product Code. 16192854
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16192854 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16192854 Supplier Abnova Supplier No. H00008625AP22.1Set
Includes
Capture antibody: rabbit MaxPab affinity purified polyclonal anti-RFXANK (100μg) , Detection antibody: mouse purified polyclonal anti-RFXANK (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Specifications
Antigen RFXANK
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RFXANK.
Gene RFXANK
Gene Alias ANKRA1/BLS/F14150_1/MGC138628/RFX-B
Gene Symbols RFXANK
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8625
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-RFXANK (100 μg) 2. Detection antibody: mouse purified polyclonal anti-RFXANK (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid