missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Rat CD43 (W3/13), Proteintech
Mouse Monoclonal Antibody
72.00€ - 222.00€
Specifications
| Antigen | CD43 |
|---|---|
| Clone | W3/13 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234448
|
Proteintech
65237-1-IG-100UG |
100 μg |
84.00€ 72.00€ / 100µg Save 12.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236601
|
Proteintech
65237-1-IG-500UG |
500 μg |
258.00€ 222.00€ / 500µg Save 36.00€ 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD43 antibody from Proteintech is a mouse monoclonal antibody generated with rat thymocyte membrane glycoproteins of rat CD43. This antibody recognizes rat antigen. The CD43 antibody has been validated for the following applications: FC analysis.Specifications
| CD43 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 24796 | |
| IgG1 κ | |
| Affinity purification | |
| Flow Antibody |
| W3/13 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Rat | |
| Rat thymocyte membrane glycoproteins | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title