missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA7 Antibody (7A1-144), Novus Biologicals™
Rat Monoclonal Antibody
165.00€ - 481.00€
Specifications
| Antigen | ABCA7 |
|---|---|
| Clone | 7A1-144 |
| Concentration | 1.3 mg/mL |
| Dilution | Western Blot 1:2000, Flow Cytometry 1:500 - 1:4000, Immunohistochemistry-Paraffin 1:100 - 1:250 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18735303
|
Novus Biologicals
NB400-163SS |
0.025 mL |
165.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18708093
|
Novus Biologicals
NB400-163 |
0.1 mL |
481.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABCA7 Monoclonal specifically detects ABCA7 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ABCA7 | |
| 1.3 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| ABC Transporters, Astrocyte Markers, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| ABCA-SSN, ABCXATP-binding cassette sub-family A member 7, ATP-binding cassette, sub-family A (ABC1), member 7, Autoantigen SS-N, EC 2.2.1.1, EC 3.6.3, EC 3.6.3.41, FLJ40025, Macrophage ABC transporter | |
| ABCA7 | |
| IgG2a κ | |
| Protein G purified | |
| 240 kDa |
| 7A1-144 | |
| Western Blot 1:2000, Flow Cytometry 1:500 - 1:4000, Immunohistochemistry-Paraffin 1:100 - 1:250 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 10347 | |
| Mouse ABCA7 stably-transfected HeLa cells. [UniProt# Q91V24] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title