missing translation for 'onlineSavingsMsg'
Learn More

Ras-GAP Mouse, Unlabeled, Clone: 13, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610040

Product Code. 15895219

  • 176.00€ / 1 set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ras-GAP, a protein of approximately 120kDa, is thought to serve as a down-regulator of p21ras by stimulating its otherwise weak intrinsic GTPase activity. Both growth factor receptor and oncogene-encoded tyrosine kinases require functional membrane-associ

Host Species: Mouse
Clone: 13
Isotype: IgG1
Species Reactivity: Canine
Immunogen: Human Ras-GAP aa.381 to 390
Formula Weight [Chemical]: 120kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ras-GAP
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Ras-GAP aa.381 to 390
50 μg
Cell Biology
Canine, Chicken, Frog, Human, Mouse, Rat
IgG1
Western Blot
13
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.