missing translation for 'onlineSavingsMsg'
Learn More

Anti-Ran antibody, Abcam

Product Code. 32107319 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Unit Size:
1mL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32107319 1 mL 1mL
32104654 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32107319 Supplier Abcam Supplier No. ab311181mL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Anti-Ran antibody

The product is subject to the following: Abcam Restricted Use Statement

.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen RAN
Applications Immunohistochemistry (FFPE)
Cellular Localization Nucleus; Nucleus envelope; Cytoplasm; Cytosol; Cytoplasm; Melanosome; Predominantly nuclear during interphase (PubMed:10679025, PubMed:12194828, PubMed:8421051). Becomes dispersed throughout the cytoplasm during mitosis (PubMed:12194828, PubMed:8421051). Identified by mass spectrometry in melanosome fractions from stage I to stage IV (PubMed:17081065).
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.6, Preservative: 0.1% Sodium azide, Constituents: PBS, 1% BSA
Gene RAN
Gene Accession No. P62826
Host Species Rabbit
Immunogen RAN
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Metabolism
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene RAN
Target Molecule RAN
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.