Learn More
PTRF, Human, IP-WB Antibody Pair (PW3), Abnova™
Product Code. 16183914
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16183914 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16183914 Supplier Abnova Supplier No. H00284119PW3.1Set
Includes
Antibody pair for IP: rabbit polyclonal anti-PTRF (300μL), Antibody pair for WB: mouse purified polyclonal anti-PTRF (50μg)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen PTRF
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene PTRF
Gene Alias FKSG13
Gene Symbols PTRF
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284119
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-PTRF (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-PTRF (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid