missing translation for 'onlineSavingsMsg'
Learn More

PTH, Human, IP-WB Antibody Pair (PW4), Abnova™

Product Code. 16142514
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16142514 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16142514 Supplier Abnova Supplier No. H00005741PW4.1Set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Antibody pair for IP: mouse monoclonal anti-PTH (300μg), Antibody pair for WB: rabbit polyclonal anti-PTH (50μL)

For immunoprecipitation and to detect the precipitated protein in western blot.

Specifications

Antigen PTH
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene PTH
Gene Alias PTH1
Gene Symbols PTH
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5741
Includes Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-PTH (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-PTH (50 ul)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less

For Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.