missing translation for 'onlineSavingsMsg'
Learn More

Anti-Prothrombin antibody [RM1252], Abcam

Product Code. 32070880 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
Unit Size:
100µL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32070880 100 μL 100µL
32070103 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32070880 Supplier Abcam Supplier No. ab322113100uL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Polyclonal Antibody

Rabbit Recombinant Multiclonal Thrombin antibody. Suitable for IHC-P, IP, WB and reacts with Human, Mouse, Rat samples.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen F2 (Thrombin)
Applications Immunohistochemistry (FFPE), Immunoprecipitation, Western Blot
Cellular Localization Secreted; Extracellular space
Classification Recombinant Polyclonal
Clone RM1252
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene F2
Gene Accession No. P00734
Host Species Rabbit
Immunogen F2 (Thrombin)
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cardiovascular
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene F2
Target Molecule F2 (Thrombin)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.